Orders LinkContactUsLinkComponent
Biotin kits, Micro HOOK™
Biotin kits, Micro HOOK™
HOOK™-NHS-dPEG4-Biotin Kit, 1 KIT
Catalog # 786-697
Supplier:   G-Biosciences
Biotin kits, Micro HOOK™
Catalog # 786-697
Supplier:  G-Biosciences
CAS Number:  
Biotin kits, Micro HOOK™
Biotin kits, Micro HOOK™
HOOK™-NHS-dPEG4-Biotin Kit, 1 KIT
CAS Number:  
Price
534,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
The micro HOOK™ Biotin kits are designed to label small amounts of proteins, with each kit designed for 8 to 10 labellings of 50 to 250 µg protein/reaction.
  • Micro kit for labelling protein primary amines
  • Optimizer Buffer™ for improved coupling efficiency
  • Gel filtration columns for rapid (<10 minute) purification
  • Labels 50 to 250µg protein/reaction
  • Suitable for 8 to 10 couplings
Kits contain the following Biotin reagents
HOOK™ Sulfo-NHS-Biotin - amine reactive reagent, shortest spacer arm
HOOK™ Sulfo-NHS-LC-Biotin - amine reactive reagent, longer spacer arm
HOOK™ Sulfo-NHS-SS-Biotin - cleavable, amine reactive reagent
HOOK™ NHS-dPEG₄-Biotin - amine reactive pegylated reagent
Specifications
Specifications
  • Pk:
    1 KIT
  • Description:
    HOOK™-NHS-dPEG4-Biotin Kit