Order Entry
Austria
Orders LinkContactUsLinkComponent
Specimen containers, disposable

Specimen containers, disposable

1000 ml, 140 mm, 111 mm
Catalog # SIMPC581-1L
Supplier:   Simport Scientific
Specimen containers, disposable
Catalog # SIMPC581-1L
Supplier:  Simport Scientific
Specimen containers, disposable
Specimen containers, disposable

1000 ml, 140 mm, 111 mm

Price
257,00 €
Log in for availability
1 * 100 ST
Product Details & Documents

These containers and closures are made of PP. They are ideal for collection, transport and storage of fluids, powders, solids, pathology specimens and hazardous samples.

  • Disposable
  • Resist temperatures from −20 to + 80 °C

Tight-fitting lids prevent leaks and odors. Certain models listed below have a tight-fitting lid. Also offered with an incorporate tamperproof lid which is opened by first removing tab on side of lid, then by lifting the cover.

The 2600 and 5700 ml sizes are provided with a plastic handle to facilitate handling.

Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904

Documents

Specifications
  • Ø opening:
    111 mm
  • Capacity:
    1000 ml
  • Height:
    140 mm
  • Seq:
    06