Orders LinkContactUsLinkComponent
Reference electrode filling solutions
Reference electrode filling solutions
FRISCOLYT-B®, for measurement at low temperature and for media with organic compounds (oil, proteins etc.), 250 ml, 6
Catalog # METT51350084
Supplier:   Mettler - Toledo
Reference electrode filling solutions
Catalog # METT51350084
Supplier:  Mettler - Toledo
CAS Number:  
Reference electrode filling solutions
Reference electrode filling solutions
FRISCOLYT-B®, for measurement at low temperature and for media with organic compounds (oil, proteins etc.), 250 ml, 6
CAS Number:  
Price
105,00 €
Log in for availability
1 * 6 ST
Product Details & Documents
FRISCOLYT-B® (84,4% glycerol, 11% KCl and 4,6% H₂O) is for low temperature samples.
Specifications
Specifications
  • Pk:
    6
  • Description:
    FRISCOLYT-B®, for measurement at low temperature and for media with organic compounds (oil, proteins etc.), 250 ml