Receiving flasks
3000 ml, Low temperature, KS 35/20
Catalog # BUCH040777
Supplier: BUCHI
Jump to:
Product Details & Documents
Borosilicate glass 3.3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Especially for BÜCHI rotary evaporators. With spherical joint socket KS 35/20.
Specifications
- Socket:KS 35/20
- Type:Low temperature
- Capacity:3000 ml