ContactUsLinkComponent
CoolRack 50 mL to hold up 4 x 50 mL centrifuge tubes. Maintains temperature through direct contact with ice, dry ice, water, liquid nitrogen or other common laboratory temperature sources. Use for cooling, (snap)freezing or heating/thawing samples. 1 * 1 items
Catalog # 479-0958
Supplier:  BIOCISION
CoolRack 50 mL to hold up 4 x 50 mL centrifuge tubes. Maintains temperature through direct contact with ice, dry ice, water, liquid nitrogen or other common laboratory temperature sources. Use for cooling, (snap)freezing or heating/thawing samples. 1 * 1 items
Catalog # 479-0958
Supplier:  BIOCISION
Frequently Bought Together