ContactUsLinkComponent
SHAFT STIRRER ANCHOR PTFE 1.6X75X18CM 1 * 1 items
Catalog # COWI005.0375.16.12
Supplier:  COWIE
SHAFT STIRRER ANCHOR PTFE 1.6X75X18CM 1 * 1 items
Catalog # COWI005.0375.16.12
Supplier:  COWIE
Frequently Bought Together