Reacti-Therm™ modules
Reacti-Therm™ module with heating function, for 27 port sample concentrator, 3, Ambient +10...200 °C
Catalog # HYPETS-18824
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Specifications
- No. of blocks:3
- Temperature Range:Ambient +10...200 °C
- Description:Reacti-Therm™ module with heating function, for 27 port sample concentrator
- Seq:014
Accessories
Product Information
- Catalog #AvailabilityPrice
- Remote temperature probe, for Reacti-Therm™ heating and stirring modulesRemote temperature probe, for Reacti-Therm™ heating and stirring modulesHYPETS-18820Log in for availability1 * 1 ST - 371,00 €