Orders LinkContactUsLinkComponent
Waste caps
Waste cap, includes O-ring, 1/4" nuts, ferrules and plug, DIN 60, 3 ports with 1/4-28 thread for 1/8" or 1/16" tubing, 1 port with 1/2-20 thread for 1/4" tubing, PP
Catalog # JOURJR-S-16003
Supplier:   VICI JOUR RESEARCH
Waste caps
Catalog # JOURJR-S-16003
Supplier:  VICI JOUR RESEARCH
Waste caps
Waste cap, includes O-ring, 1/4" nuts, ferrules and plug, DIN 60, 3 ports with 1/4-28 thread for 1/8" or 1/16" tubing, 1 port with 1/2-20 thread for 1/4" tubing, PP
Price
259,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
These waste caps include three or four ports for ¹/₈ and ¹/₁₆" tubing. Two versions of each waste cap are available, with an extra port to accommodate either a 1/2-20 port for ¹/₄" O.Ø tubing, or a barbed hose adapter.
 : Barbed adapters must be ordered separately.
Specifications
  • Thread:
    DIN 60
  • Description:
    Waste cap, includes O-ring, 1/4" nuts, ferrules and plug
  • Material:
    PP
  • Seq:
    08
  • Connector:
    3 ports with 1/4-28 thread for 1/8" or 1/16" tubing, 1 port with 1/2-20 thread for 1/4" tubing