ContactUsLinkComponent
Cuvette pp 14 lt grey with two handles, stackable, chemicals and hot water-resistant, temperature resistant up to +130 c. colors m ay vary from illustration reserved. 1 * 1 items
Catalog # SEMA0731
Supplier:  SEMADENI
Cuvette pp 14 lt grey with two handles, stackable, chemicals and hot water-resistant, temperature resistant up to +130 c. colors m ay vary from illustration reserved. 1 * 1 items
Catalog # SEMA0731
Supplier:  SEMADENI
Frequently Bought Together