Buffer solutions
Buffer standard DIN 19266, 4,005, -, 25 °C, Plastic bottle, 500 mL
Supplier: REAGECON
Jump to:
Product Details & Documents
Dokumente
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Plastic bottle
- Pk:500 mL
- Reference temperature:25 °C
- Description:Buffer standard DIN 19266
- pH accuracy:-
- pH value:4,005



