Orders LinkContactUsLinkComponent
Microtubes, with Thread
Microtubes, with Thread
Micro tubes with skirted base, neutral screw cap assembled, sterile, without knurls, 0,5 ml, 1000/case
Catalog # SARS72.785.005
Marketsource
Supplier:   SARSTEDT
Microtubes, with Thread
Catalog # SARS72.785.005
Supplier:  SARSTEDT
Marketsource
Microtubes, with Thread
Micro tubes with skirted base, neutral screw cap assembled, sterile, without knurls, 0,5 ml, 1000/case
Price
119,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents
PP, translucent.
  • Safe screw cap with O-ring
  • Sterile or non sterile
  • Autoclavable
For transport, storage and sample processing in the laboratory.
Specifications
  • Packed:
    1000/case
  • Capacity:
    0,5 ml
  • Description:
    Micro tubes with skirted base, neutral screw cap assembled, sterile, without knurls
  • Seq:
    17