ContactUsLinkComponent
Safety cabinet with wing door. Outer carcass: 595 x 595 x 2080 (WxDxH) mm, Inner carcass: 499 x 446 x 1830 (WxDxH) mm, Wing door hung right Outer carcass, sheet steel, powder-coated in RAL 7035 light grey, Wing door in RAL 7035 light grey, Inner carc 1 * 1 items
  132-0652
 :  DUPERTHAL
Safety cabinet with wing door. Outer carcass: 595 x 595 x 2080 (WxDxH) mm, Inner carcass: 499 x 446 x 1830 (WxDxH) mm, Wing door hung right Outer carcass, sheet steel, powder-coated in RAL 7035 light grey, Wing door in RAL 7035 light grey, Inner carc 1 * 1 items
  132-0652
 :  DUPERTHAL
Frequently Bought Together