ContactUsLinkComponent
Blade, trapezoid no.5634 length: 53.00mm, width: 19.00mm, material thickness: 0.63mm, non-corrosive Blade, 2-sided ground-edge, doub le bevel edge-ground, 2-times usable Blade. 1 * 10 items
Catalog # MARO610.70
Supplier:  MARTOR ARGENTAX
Blade, trapezoid no.5634 length: 53.00mm, width: 19.00mm, material thickness: 0.63mm, non-corrosive Blade, 2-sided ground-edge, doub le bevel edge-ground, 2-times usable Blade. 1 * 10 items
Catalog # MARO610.70
Supplier:  MARTOR ARGENTAX
Frequently Bought Together