ContactUsLinkComponent
Screw fitting,swiveling,for sensor/tube outer diameter: 3.2mm,for thread : GL 18, maximum angle : 9°, fix all kinds of sensors (stai nless steel, glass, PTFE), tubes and tight tubing in the requested angle,suitable for both pressure and vacuum. 1 * 1 items
Catalog # BOHLD690-18
Supplier:  Bohlender
undefined
Screw fitting,swiveling,for sensor/tube outer diameter: 3.2mm,for thread : GL 18, maximum angle : 9°, fix all kinds of sensors (stai nless steel, glass, PTFE), tubes and tight tubing in the requested angle,suitable for both pressure and vacuum. 1 * 1 items
Catalog # BOHLD690-18
Supplier:  Bohlender
Frequently Bought Together