Orders LinkContactUsLinkComponent
Sample vials, closed-top
Sample vials, closed-top
20,00 ml, Clear, Yes, Closed, 24-414, PTFE-faced
Catalog # EPSCC126-0020
   Low Manufacturing Impact
Supplier:   EP SCIENTIFIC
Sample vials, closed-top
Catalog # EPSCC126-0020
Supplier:  EP SCIENTIFIC
   Low Manufacturing Impact
Sample vials, closed-top
Sample vials, closed-top
20,00 ml, Clear, Yes, Closed, 24-414, PTFE-faced
Price
185,00 €
Log in for availability
1 * 72 ST
Product Details & Documents
Clear or amber sample vials made from borosilicate glass.
  • Designed for water sample collection for volatile organic analysis (VOA)
  • Optimised for use with automation
  • With closed-top white PP cap bonded with PTFE-lined silicone septum for secure sealing
Processed or certified to meet or exceed EPA Performance Based Specifications for volatile organic analysis with certificate. Processed vials are processed in the same manner as certified vials and come with all the features of certified vials, but vials are not barcoded and CoA is not included.
Specifications
  • Cap included:
    Yes
  • Cap Size:
    24-414
  • Septum:
    PTFE-faced
  • Capacity:
    20,00 ml
  • Colour:
    Clear
  • Seq:
    01
  • Cap type:
    Closed
  • Cat. No.:
    EPSCC126-0020
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact