ContactUsLinkComponent
Magnetic stirrer, wear-free with 1 stirring position, for up to 10,000 ml stirring volume.1 stirring position, 100% maintenance-free and wear-free by inductive 2mag- 1 * 1 items
Catalog # TMAG40001_UK
Supplier:  2MAG
Magnetic stirrer, wear-free with 1 stirring position, for up to 10,000 ml stirring volume.1 stirring position, 100% maintenance-free and wear-free by inductive 2mag- 1 * 1 items
Catalog # TMAG40001_UK
Supplier:  2MAG
Frequently Bought Together