Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Capillary tubes

150 mm, 1,00 mm, 0,80 mm
  HILG1415017
 :   HILGENBERG
Capillary tubes
  HILG1415017
Capillary tubes
Capillary tubes

150 mm, 1,00 mm, 0,80 mm

 
622,00 €
   
1 * 1.000 ST
 

Soda-lime glass.

  • Both ends open
 
  • Ext.Ø:
    1,00 mm
  • Ø int.:
    0,80 mm
  • Length:
    150 mm
  • Seq:
    11
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.