WASTE BOX WITH BAG 57L 1 * 25 items
SCAA95001159-1
: Essity
Frequently Bought Together
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Detergents and cleaning agents, LABWASH® Premium
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Pipette tips, Finntip™
: Thermo Fisher Scientific
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Single-use syringes, 3-piece, HENKE-JECT®
: HENKE SASS WOLF