Wire baskets, rectangular, with lid
274×174×36 mm
HUPF7501135
These fine mesh baskets, made of stainless steel, electropolished, are designed to keep small parts and accessories.
- With lid
- With base frame
- Mesh size: 1,6×1,6 mm
Colour: Silver.
Documents
HUPF7501135_TDS_EN
Datasheet
- Seq:06
- W×D×H:274×174×36 mm



