Orders LinkContactUsLinkComponent
Bucket, Rectangular
Bucket, Rectangular
Bucket with colour coding clips, Grey, 6 L
Catalog # 129-0458
Supplier:   FHP VILEDA SCS
Bucket, Rectangular
Catalog # 129-0458
Supplier:  FHP VILEDA SCS
Bucket, Rectangular
Bucket, Rectangular
Bucket with colour coding clips, Grey, 6 L
Price
25,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Polypropylene bucket with stainless steel handle. For use on the CE Bucket Trolley or the CE pre-prepared trolley.
  • Liquid measurement lines molded inside of bucket
  • Color coding clips included to allow differentiation in colours of buckets (red, blue, green, yellow)
  • Autoclavable
Highest performance for the best contamination control.
Specifications
  • Capacity:
    6 L
  • Colour:
    Grey
  • Description:
    Bucket with colour coding clips
  • Seq:
    01