Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
FREEZING POINT STD CRYOSCOPE 621 100ML 1 * 1 items
Catalog # REGEMTR040X
Supplier:
REAGECON
Certificates
Expand image
FREEZING POINT STD CRYOSCOPE 621 100ML 1 * 1 items
Catalog # REGEMTR040X
Supplier:
REAGECON
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
Documentation
Frequently Bought Together
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
Supplier: Merck
Potassium chloride 3 mol/l (3 N) in aqueous solution, Supelco®
Supplier: Merck
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Nitrate standard solution, 1000 mg/L NO₃<sup>-</sup> in Water (from NaNO₃) Certipur®, Supelco®
Supplier: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Magnetic Stirring Bars, Plain, Economy
Supplier: VWR Collection
FREEZING POINT STD CRYOSCOPE 422 100ML 1 * 1 items
Supplier: REAGECON
VWR® Culture Media, Ready To Use, In Plates
Supplier: VWR Chemicals