Ring weights for flasks
Viny coated lead ring weight, Erlenmeyer 500 - 2000 ml, 57 mm
Catalog # DELT192472
Supplier: DELTALAB
Springe zu:
Product Details & Documents
Closed flask weight rings with heavy vinyl coating, which is ideal to improve the stability of Erlenmeyer flasks in a water bath.
- High chemical resistance
Spezifikationen
- Description:Viny coated lead ring weight
- Seq:03
- For:Erlenmeyer 500 - 2000 ml
- Ø int.:57 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



