Order Entry
Austria
Orders LinkContactUsLinkComponent
Ring weights for flasks

Ring weights for flasks

Viny coated lead ring weight, Erlenmeyer 125 - 500 ml, 48 mm
  DELT192470
 :   DELTALAB
Ring weights for flasks
  DELT192470
 :  DELTALAB
Ring weights for flasks

Viny coated lead ring weight, Erlenmeyer 125 - 500 ml, 48 mm

 
182,00 €
   
1 * 1 ST
 

Closed flask weight rings with heavy vinyl coating, which is ideal to improve the stability of Erlenmeyer flasks in a water bath.

  • High chemical resistance
 
  • Description:
    Viny coated lead ring weight
  • Seq:
    01
  • For:
    Erlenmeyer 125 - 500 ml
  • Ø int.:
    48 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.