Orders LinkContactUsLinkComponent
Interfaces
Interfaces
Com.G@te®, internal for Petite Fleur®, Grande Fleur®, Unichiller® with Pilot ONE, Ministats®, CC®-300BX to CC®-906w
Catalog # HUBE31217
Supplier:   HUBER
Interfaces
Catalog # HUBE31217
Supplier:  HUBER
Interfaces
Interfaces
Com.G@te®, internal for Petite Fleur®, Grande Fleur®, Unichiller® with Pilot ONE, Ministats®, CC®-300BX to CC®-906w
Price
715,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Com.G@te®. The interfaces are integrated RS232 (bi-directional), RS485 (bi-directional), ECS external control signal, volt free contact, AIF analogue interface 0/4 to 20 mA or 0 to 10 V (bi-directional). Connections comply with NAMUR standard.
Specifications
  • Description:
    Com.G@te®, internal for Petite Fleur®, Grande Fleur®, Unichiller® with Pilot ONE, Ministats®, CC®-300BX to CC®-906w
  • Seq:
    01