ContactUsLinkComponent
Areometer, large tub, plato type, measuring range:20-30:0.1%mas, without thermometer, reading above, length:650mm, reference tempera ture: 6°C. 1 * 1 items
Catalog # AMARH847304
Supplier:  Amarell
Areometer, large tub, plato type, measuring range:20-30:0.1%mas, without thermometer, reading above, length:650mm, reference tempera ture: 6°C. 1 * 1 items
Catalog # AMARH847304
Supplier:  Amarell
Frequently Bought Together