Orders LinkContactUsLinkComponent
Filter sets
Filter set 63 (high efficiency), excitation BP 572/25 (HE), beam splitter FT 590 (HE), emission BP 629/62 (HE)
Catalog # ZEIS4890630000000
Supplier:   ZEISS
Filter sets
Catalog # ZEIS4890630000000
Supplier:  ZEISS
Filter sets
Filter set 63 (high efficiency), excitation BP 572/25 (HE), beam splitter FT 590 (HE), emission BP 629/62 (HE)
Price
Inquire For Price
Log in for availability
1 * 1 SET
Product Details & Documents
Specifications
  • Description:
    Filter set 63 (high efficiency), excitation BP 572/25 (HE), beam splitter FT 590 (HE), emission BP 629/62 (HE)
  • Seq:
    13