Orders LinkContactUsLinkComponent
VWR® Basic Frocks, SMS
VWR® Basic Frocks, SMS
Blue, L
Catalog # 113-1174
Supplier:   VWR Collection
VWR® Basic Frocks, SMS
Catalog # 113-1174
Supplier:  VWR Collection
VWR® Basic Frocks, SMS
VWR® Basic Frocks, SMS
Blue, L
Price
200,00 €
Log in for availability
1 * 30 ST
Product Details & Documents
These SMS frocks are manufactured from a latex-free, multi-layer spunbonded - melt-blown - spunbonded (SMS) fabric, that offers high tensile strength while remaining soft, comfortable and breathable.
  • Snap collar, raglan sleeves, snap closure
  • Fluid and particulate barrier
  • Light chemical splash resistance
Colour: White or blue
Specifications
  • Clothing Size:
    L
  • Colour:
    Blue
  • Seq:
    08
  • Cat. No.:
    113-1174
  • Garment application:
    Cleanroom|Laboratory
  • Gender:
    Unisex
  • Pockets:
    Without pockets
  • Closure type:
    Press studs / snaps