Orders LinkContactUsLinkComponent
VWR® Basic Frocks, SPP
VWR® Basic Frocks, SPP
White, 3XL
Catalog # 113-1155
Supplier:   VWR Collection
VWR® Basic Frocks, SPP
Catalog # 113-1155
Supplier:  VWR Collection
VWR® Basic Frocks, SPP
VWR® Basic Frocks, SPP
White, 3XL
Price
116,00 €
Log in for availability
1 * 30 ST
Product Details & Documents
VWR® Basic SPP frocks are manufactured from a strong, lightweight and breathable, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
  • Snap collar, raglan sleeves and snap closure
  • Elasticated cuffs, without pockets
  • Lightweight and breathable first line barrier protection
This garment is ideal for less critical areas or pre-gowning entry rooms. It is tested and manufactured in an ISO certified facility that is lot controlled and validated through independent laboratory testing.
Specifications
  • Clothing Size:
    3XL
  • Colour:
    White
  • Cat. No.:
    113-1155
  • Garment application:
    Cleanroom|Laboratory
  • Gender:
    Unisex
  • Pockets:
    Without pockets
  • Closure type:
    Press studs / snaps
  • Weight:
    48 g/m²