Tape autoclave 19 mm x 55 m self-adhesive tape for easy removability proper control of the process of autoclaving and temperature. a fter reaching 121 ° c, the indication color stripes in a dark brown. tape length 50 m, width 19 mm 1 * 1 Roll
Catalog # SEMA4343
Supplier: SEMADENI
Frequently Bought Together