Slide box
Slide box, Red, Cardboard, 50
Catalog # HECH42705003
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
Stackable storage boxes with tight-closing lid, removable index card and numbered bottom.
Specifications
- No. of slides:50
- Colour:Red
- Description:Slide box
- Material:Cardboard