Order Entry
Austria
Orders LinkContactUsLinkComponent
Buffer solutions

Buffer solutions

High Resolution Colour Coded Buffer, 7,000, -, Yellow, 25 °C, Plastic bottle, 500 mL
Catalog # REGEH70525C
Supplier:   REAGECON
Buffer solutions
Catalog # REGEH70525C
Supplier:  REAGECON
CAS Number:  
Buffer solutions
Buffer solutions

High Resolution Colour Coded Buffer, 7,000, -, Yellow, 25 °C, Plastic bottle, 500 mL

CAS Number:  
Price
42,10 €
Log in for availability
1 * 500 mL
Product Details & Documents

Documents

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Reference temperature:
    25 °C
  • Colour:
    Yellow
  • Description:
    High Resolution Colour Coded Buffer
  • pH accuracy:
    -
  • pH value:
    7,000

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.