Orders LinkContactUsLinkComponent
Sample storage tubes, blank and alphanumeric
Sample storage tubes, blank and alphanumeric
1,2 ml, Microtubes in a cluster rack with lid, each rack containing 96 microtubes, 10 racks of 96 tubes/case, Blank storage tube
Artikelnr. 732-4827
   Low Manufacturing Impact
Sample storage tubes, blank and alphanumeric
Artikelnr. 732-4827
   Low Manufacturing Impact
Sample storage tubes, blank and alphanumeric
Sample storage tubes, blank and alphanumeric
1,2 ml, Microtubes in a cluster rack with lid, each rack containing 96 microtubes, 10 racks of 96 tubes/case, Blank storage tube
Preis
544,00 €
Log in for availability
1 * 960 ST
Product Details & Documents
Abgene blank and alphanumeric storage tubes are manufactured from 100% virgin, medical-grade polypropylene. Storage tubes are leak tested to ensure the integrity and security of samples and with a V-bottom design, our alphanumeric storage tubes facilitate sample recovery.
  • Highest quality
  • Enhanced rack design
Unlike traditional storage tubes or blocks, our blank and alphanumeric storage tubes are available in our specially-designed, stackable, 96-format, microplate-footprint racks to save precious space in your freezer, refrigerator, compound store or on the bench top. In addition, the rack design includes grid referenced lids for simple identification of tube location.
Specifications
  • Packed:
    10 racks of 96 tubes/case
  • Type:
    Blank storage tube
  • Capacity:
    1,2 ml
  • Description:
    Microtubes in a cluster rack with lid, each rack containing 96 microtubes
  • Seq:
    02
  • Cat. No.:
    732-4827
  • Environmental certification:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Environmentally Preferable:
    Low Manufacturing Impact