Orders LinkContactUsLinkComponent
Microsampling tools
Microsampling tools
Aluminum probe handle
Catalog # PERCN9302622
Supplier:   PerkinElmer
Microsampling tools
Catalog # PERCN9302622
Supplier:  PerkinElmer
Microsampling tools
Aluminum probe handle
Price
45,30 €
Log in for availability
1 * 1 ST
Product Details & Documents
This microsampling tool kit is suitable for FTIR microscopy.
Delivery information: Microsampling tool kit includes dissecting forceps, roller knife, straight micro probe, aluminum probe handle, pin vise.
Specifications
  • Description:
    Aluminum probe handle
  • Seq:
    05