Microsampling tools
Micro specimen tweezers
Artikelnr. PERCN9302609
Anbieter: PerkinElmer
Springe zu:
Product Details & Documents
This microsampling tool kit is suitable for FTIR microscopy.
Delivery information: Microsampling tool kit includes dissecting forceps, roller knife, straight micro probe, aluminum probe handle, pin vise.
Spezifikationen
- Description:Micro specimen tweezers
- Seq:03