Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Viscosity reference standards

Nominal viscosity: 930 cSt at 40 °C and 82 cSt at 100 °C, 500 ml, Hydrocarbons, 1
Catalog # PERCN5316005
Supplier:   PerkinElmer
Viscosity reference standards
Catalog # PERCN5316005
Supplier:  PerkinElmer
CAS Number:  
Viscosity reference standards
Viscosity reference standards

Nominal viscosity: 930 cSt at 40 °C and 82 cSt at 100 °C, 500 ml, Hydrocarbons, 1

CAS Number:  
Price
169,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications

Specifications

  • Pk:
    1
  • Description:
    Nominal viscosity: 930 cSt at 40 °C and 82 cSt at 100 °C, 500 ml
  • Type:
    Hydrocarbons

Specifications