Viscosity reference standards
Nominal viscosity: 360 cSt at 40 °C and 42 cSt at 100 °C, 500 ml, Hydrocarbons, 1
Catalog # PERCN5316004
Supplier: PerkinElmer
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:1
- Description:Nominal viscosity: 360 cSt at 40 °C and 42 cSt at 100 °C, 500 ml
- Type:Hydrocarbons
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



