Order Entry
Austria
Orders LinkContactUsLinkComponent
Viscosity reference standards

Viscosity reference standards

Nominal viscosity: 100 cSt at 40 °C and 16,8 cSt at 100 °C, 500 ml, Hydrocarbons, 1
Catalog # PERCN5316003
Supplier:   PerkinElmer
Viscosity reference standards
Catalog # PERCN5316003
Supplier:  PerkinElmer
CAS Number:  
Viscosity reference standards
Viscosity reference standards

Nominal viscosity: 100 cSt at 40 °C and 16,8 cSt at 100 °C, 500 ml, Hydrocarbons, 1

CAS Number:  
Price
169,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications

Specifications

  • Pk:
    1
  • Description:
    Nominal viscosity: 100 cSt at 40 °C and 16,8 cSt at 100 °C, 500 ml
  • Type:
    Hydrocarbons

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.