Order Entry
Austria
Orders LinkContactUsLinkComponent
Viscosity reference standards

Viscosity reference standards

Viscosity standard reference kit, 5×500 ml, Hydrocarbons, 1
Catalog # PERCN5316000
Supplier:   PerkinElmer
Viscosity reference standards
Catalog # PERCN5316000
Supplier:  PerkinElmer
CAS Number:  
Viscosity reference standards
Viscosity reference standards

Viscosity standard reference kit, 5×500 ml, Hydrocarbons, 1

CAS Number:  
Price
939,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications

Specifications

  • Pk:
    1
  • Description:
    Viscosity standard reference kit, 5×500 ml
  • Type:
    Hydrocarbons

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.