Rectangular spacers for SpectrumGX
Assorted spacers for semi-demountable cell, includes two each of thickness 0,025, 0,05, 0,1, 0,2, 0,5, 1,0 mm spacers, SpectrumGX
Catalog # PERCL1270421
Supplier: PerkinElmer
Jump to:
Product Details & Documents
PTFE spacers can be used for the analysis of less viscous samples.
Specifications
- For:SpectrumGX
- Description:Assorted spacers for semi-demountable cell, includes two each of thickness 0,025, 0,05, 0,1, 0,2, 0,5, 1,0 mm spacers
- Seq:07