Orders LinkContactUsLinkComponent
Rectangular spacers for SpectrumGX
Rectangular spacers for SpectrumGX
Spacers, PTFE, rectangular, 0,2 mm thickness for semi-demountable cell, SpectrumGX
Catalog # PERCL1270408
Supplier:   PerkinElmer
Rectangular spacers for SpectrumGX
Catalog # PERCL1270408
Supplier:  PerkinElmer
Rectangular spacers for SpectrumGX
Spacers, PTFE, rectangular, 0,2 mm thickness for semi-demountable cell, SpectrumGX
Price
218,00 €
Log in for availability
1 * 3 ST
Product Details & Documents
PTFE spacers can be used for the analysis of less viscous samples.
Specifications
  • For:
    SpectrumGX
  • Description:
    Spacers, PTFE, rectangular, 0,2 mm thickness for semi-demountable cell
  • Seq:
    04