Rectangular spacers for SpectrumGX
Spacers, PTFE, rectangular, 0,05 mm thickness for semi-demountable cell, SpectrumGX
Catalog # PERCL1270402
Supplier: PerkinElmer
Jump to:
Product Details & Documents
PTFE spacers can be used for the analysis of less viscous samples.
Specifications
- For:SpectrumGX
- Description:Spacers, PTFE, rectangular, 0,05 mm thickness for semi-demountable cell
- Seq:02