Orders LinkContactUsLinkComponent
Microsampling tools
Microsampling tools
Microsampling tool kit
Catalog # PERCN1870151
Supplier:   PerkinElmer
Microsampling tools
Catalog # PERCN1870151
Supplier:  PerkinElmer
Microsampling tools
Microsampling tool kit
Price
737,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
This microsampling tool kit is suitable for FTIR microscopy.
Delivery information: Microsampling tool kit includes dissecting forceps, roller knife, straight micro probe, aluminum probe handle, pin vise.
Specifications
  • Description:
    Microsampling tool kit
  • Seq:
    01