Orders LinkContactUsLinkComponent
Calibration weight sets, class E2, Proof Line
Calibration weight sets, class E2, Proof Line
1 g - 200 g, With DAkkS certificate
Catalog # SARBYCS31-522-AC02
Supplier:   Sartorius Balances
Calibration weight sets, class E2, Proof Line
Catalog # SARBYCS31-522-AC02
Supplier:  Sartorius Balances
Calibration weight sets, class E2, Proof Line
Calibration weight sets, class E2, Proof Line
1 g - 200 g, With DAkkS certificate
Price
1 740,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
These class E2 weight sets include aluminium (1 to 5 mg) or nickel silver (10 to 500 mg) or stainless steel (1 g and higher) weights.
Specifications
  • Weight set:
    1 g - 200 g
  • Seq:
    08
  • Type:
    With DAkkS certificate