ContactUsLinkComponent
GPRT 1400AN, DIGITAL-pH-/mV-/Thermometer Measuring range (pH): 0,00 ... 14,00 pH Measuring range (redox): -1999 ... +1999 mV Measuring range (temperature): -20,0 ... +110,0 C Analog output: 1mV/digit Temperature compensation: manually or automatical 1 * 1 items
Catalog # GRSG102425
Supplier:  Greisinger
GPRT 1400AN, DIGITAL-pH-/mV-/Thermometer Measuring range (pH): 0,00 ... 14,00 pH Measuring range (redox): -1999 ... +1999 mV Measuring range (temperature): -20,0 ... +110,0 C Analog output: 1mV/digit Temperature compensation: manually or automatical 1 * 1 items
Catalog # GRSG102425
Supplier:  Greisinger
Frequently Bought Together