Nadifloxacin certified 1 * 0,1 g
Catalog # EHERC15400500
Supplier: EHRENSTORFER
Frequently Bought Together
pH buffer solutions, certified (DKD)
Supplier: Mettler - Toledo
VWR®, General purpose viscosity standards
Supplier: VWR Chemicals
VWR®, Silicone Rotational viscosity standards
Supplier: VWR Chemicals
VWR®, Cold Cranking Simulator (CCS) viscosity standards
Supplier: VWR Chemicals
Geraniol, Supelco®
Supplier: Merck
Supelco® Organic Reference Standard, Tridecanoic acid
Supplier: Merck
Methyl ethyl ketone
Supplier: EHRENSTORFER
Palladium standard solution, 1000 mg/L Pd in 20% hydrochloric acid (from Pd), standard for ICP, Specpure®
Supplier: Thermo Fisher Scientific
Sodium Standard for AAS 1 * 250 mL
Supplier: Merck
Ruthenium standard solution, 1000 mg/L Ru in 20% hydrochloric acid (from RuCl₃), standard for ICP, Specpure®
Supplier: Thermo Fisher Scientific
Absorbent rolls for chemical spillages
Supplier: asecos
Supelco® Organic Reference Standard, (−)-Isopulegol
Supplier: Merck
Amoxicillin Related Compounds, Reference Standard
Supplier: USP
VWR®, Turbidity standards, formazin
Supplier: VWR Chemicals
Organic reference standard, Chloroform 5000 µg/ml in methanol, Dr Ehrenstorfer
Supplier: EHRENSTORFER