Order Entry
Austria
Orders LinkContactUsLinkComponent
Capillary tubes

Capillary tubes

75 mm, 2 mm, 1,6
Catalog # HILG1422037
Supplier:   HILGENBERG
Capillary tubes
Catalog # HILG1422037
Supplier:  HILGENBERG
Capillary tubes
Capillary tubes

75 mm, 2 mm, 1,6

Price
916,00 €
Log in for availability
1 * 100 ST
Product Details & Documents

Borosilicate glass, ends cut, with filament.

Specifications
  • Ext.Ø:
    2 mm
  • Ø int.:
    1,6
  • Length:
    75 mm
  • Seq:
    06
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.