Orders LinkContactUsLinkComponent
Accessories
Accessories
Dispensing cartridges, piston in PE, cylinder in PP, 2 ml, non sterile
Catalog # 613-1650
Supplier:   Brand
Accessories
Catalog # 613-1650
Supplier:  Brand
Accessories
Accessories
Dispensing cartridges, piston in PE, cylinder in PP, 2 ml, non sterile
Price
29,20 €
Log in for availability
1 * 3 ST
Specifications
  • Description:
    Dispensing cartridges, piston in PE, cylinder in PP, 2 ml, non sterile
  • Seq:
    08