Order Entry
Austria
Orders LinkContactUsLinkComponent
Volumetric flasks, class B

Volumetric flasks, class B

10 ml, 10/19, 90 mm
Artikelnr. HIRS7550160
Anbieter:   HIRSCHMANN
Volumetric flasks, class B
Artikelnr. HIRS7550160
Anbieter:  HIRSCHMANN
Volumetric flasks, class B
Volumetric flasks, class B

10 ml, 10/19, 90 mm

Preis
21,90 €
Log in for availability
1 * 2 ST
Product Details & Documents

PP, translucent, with PP NS-stopper.

Specifications
  • NS:
    10/19
  • Capacity:
    10 ml
  • Height:
    90 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.