Orders LinkContactUsLinkComponent
Tank liners, HyPerforma™
Tank liners, HyPerforma™
Tank liners, 50 l
Catalog # HYCPSH30646.01
Tank liners, HyPerforma™
Catalog # HYCPSH30646.01
Tank liners, HyPerforma™
Tank liners, 50 l
Price
2 620,00 €
Log in for availability
1 * 10 ST
Product Details & Documents
Tank liners are gamma irradiated, open single use bags designed to fit in cylindrical drums.
  • Gusset design allows for a close fit to the tank
  • Constructed of CX3-9 film
  • For HyPerforma™ Single-Use Mixer DS 300
Specifications
  • Description:
    Tank liners, 50 l
  • Seq:
    01