Order Entry
Austria
Orders LinkContactUsLinkComponent
Tank liners, HyPerforma™

Tank liners, HyPerforma™

Tank liners, with liner Ø 630 mm, 50 l
Catalog # HYCPSH30647.04
Tank liners, HyPerforma™
Catalog # HYCPSH30647.04
Tank liners, HyPerforma™

Tank liners, with liner Ø 630 mm, 50 l

Price
1 970,00 €
Log in for availability
1 * 36 ST
Product Details & Documents

Tank liners are gamma irradiated, open single use bags designed to fit in cylindrical drums.

  • Gusset design allows for a close fit to the tank
  • Constructed of CX3-9 film
  • For HyPerforma™ Single-Use Mixer DS 300
Specifications
  • Description:
    Tank liners, with liner Ø 630 mm, 50 l
  • Seq:
    05
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.